Learn More
MSC Baldwin Outer Air Element, 15 1/2 In Length, Attached Gasket
Catalog No. 5000508400
Encompass
Baldwin air compressor engine filters are designed to clean air effectively before it reaches the compressor engine, ensuring optimal performance and longevity.
  • Outer air element product type.
  • 13 13/16 in outside diameter.
  • 15/16 in and 9 1/2 in inside diameter.
  • 15 1/2 in length.
  • Attached gasket.
  • Furnished gasket included.
Catalog No. 50-005-08400 Supplier MSC Supplier No. 55986343
Baldwin air compressor engine filters are designed to clean air effectively before it reaches the compressor engine, ensuring optimal performance and longevity.
  • Outer air element product type.
  • 13 13/16 in outside diameter.
  • 15/16 in and 9 1/2 in inside diameter.
  • 15 1/2 in length.
  • Attached gasket.
  • Furnished gasket included.