Learn More
MSC Baldwin Outer Air Element, 6 3/32 in OD, 9 5/8 in Length, Attached Gasket
Catalog No. 5000508964
Encompass
Baldwin axial seal air filter elements ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. This outer air element is designed to protect your equipment from harmful particles.
  • Ensures proper equipment function
  • Prevents airborne contaminants from reaching the combustion chamber
  • Axial seal design
  • Attached gasket for secure fit
  • Suitable for Kubota Tractors
Catalog No. 50-005-08964 Supplier MSC Supplier No. 55865497
Baldwin axial seal air filter elements ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. This outer air element is designed to protect your equipment from harmful particles.
  • Ensures proper equipment function
  • Prevents airborne contaminants from reaching the combustion chamber
  • Axial seal design
  • Attached gasket for secure fit
  • Suitable for Kubota Tractors