Learn More
MSC Baldwin Outer Air Element, 8 1/2 in OD, 16 3/4 in Length, for Komatsu Equipment
Catalog No. 5000509936
Encompass
The Baldwin PA3609 is an axial seal outer air element specifically designed for Komatsu equipment. This air filter element features an outside diameter of 8 1/2 in (215.9 mm) and a length of 16 3/4 in (425.5 mm). Please note that this product is currently obsolete, with availability limited to dealer stock.
  • Axial seal air filter element
  • Outside diameter of 8 1/2 in (215.9 mm)
  • Inside diameter of 5/8 in (15.9 mm) and 4 5/32 in (105.6 mm)
  • Length of 16 3/4 in (425.5 mm)
  • Includes attached and furnished gaskets
  • Optimized for use with Komatsu equipment
Catalog No. 50-005-09936 Supplier MSC Supplier No. 55984462
The Baldwin PA3609 is an axial seal outer air element specifically designed for Komatsu equipment. This air filter element features an outside diameter of 8 1/2 in (215.9 mm) and a length of 16 3/4 in (425.5 mm). Please note that this product is currently obsolete, with availability limited to dealer stock.
  • Axial seal air filter element
  • Outside diameter of 8 1/2 in (215.9 mm)
  • Inside diameter of 5/8 in (15.9 mm) and 4 5/32 in (105.6 mm)
  • Length of 16 3/4 in (425.5 mm)
  • Includes attached and furnished gaskets
  • Optimized for use with Komatsu equipment