Learn More
MSC Baldwin Radial Seal Inner Air Element, 5.25 in OD, 3.72 in ID, 15.72 in Length
Catalog No. 5000509896
Encompass
This radial seal inner air element is designed to ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. It features a robust design to provide reliable filtration for various heavy-duty applications.
  • Prevents airborne contaminants from reaching the combustion chamber.
  • Features a radial seal design for secure fitment.
  • Functions as an inner air element.
  • Suitable for use in Mitsubishi Trucks.
Catalog No. 50-005-09896 Supplier MSC Supplier No. 55874036
This radial seal inner air element is designed to ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. It features a robust design to provide reliable filtration for various heavy-duty applications.
  • Prevents airborne contaminants from reaching the combustion chamber.
  • Features a radial seal design for secure fitment.
  • Functions as an inner air element.
  • Suitable for use in Mitsubishi Trucks.