Learn More
MSC Baldwin Radial Seal Inner Air Element, 7 5/32 in OD, 19 21/32 in Length
Catalog No. 5000508913
Encompass
Baldwin Radial Seal Air Filters are engineered to ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. These air filter elements are critical for maintaining engine health and operational efficiency.
  • Outside diameter of 7 5/32 in (181.8 mm)
  • Inside diameter of 5 27/32 in (148.4 mm)
  • Length of 19 21/32 in (499.3 mm)
  • Designed to fit with companion air filter elements
  • Suitable for specific John Deere loader models including 824K, 844J, and 844K
  • Prevents airborne contaminants from reaching the combustion chamber
Catalog No. 50-005-08913 Supplier MSC Supplier No. 55874226
Baldwin Radial Seal Air Filters are engineered to ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber. These air filter elements are critical for maintaining engine health and operational efficiency.
  • Outside diameter of 7 5/32 in (181.8 mm)
  • Inside diameter of 5 27/32 in (148.4 mm)
  • Length of 19 21/32 in (499.3 mm)
  • Designed to fit with companion air filter elements
  • Suitable for specific John Deere loader models including 824K, 844J, and 844K
  • Prevents airborne contaminants from reaching the combustion chamber