Learn More
MSC Baldwin Radial Seal Inner Air Element, 9 3/32 in OD, 15 7/32 in Length
Catalog No. 5000508345
Encompass
Baldwin Radial Seal Inner Air Elements are designed to ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber, making them suitable for demanding applications.
  • Radial seal inner air element design.
  • Outside diameter of 9 3/32 in (231.0 mm).
  • Inside diameter of 6 29/32 in (175.4 mm).
  • Length of 15 7/32 in (386.6 mm).
  • Suitable for Caterpillar equipment.
Catalog No. 50-005-08345 Supplier MSC Supplier No. 55873657
Baldwin Radial Seal Inner Air Elements are designed to ensure proper equipment function by preventing airborne contaminants from reaching the combustion chamber, making them suitable for demanding applications.
  • Radial seal inner air element design.
  • Outside diameter of 9 3/32 in (231.0 mm).
  • Inside diameter of 6 29/32 in (175.4 mm).
  • Length of 15 7/32 in (386.6 mm).
  • Suitable for Caterpillar equipment.