Learn More
MSC Baldwin Radial Seal Outer Air Element, 12.41 in OD
Catalog No. 5000508344
Encompass
The radial seal air filter element is designed to protect heavy-duty equipment by preventing airborne dirt and other contaminants from reaching the combustion chamber, ensuring proper equipment function.
  • Radial seal design utilizes a urethane compound to form molded ends and bond the media pack.
  • The design creates a strong, yet flexible package.
  • Heavy-duty radial seal gasket makes servicing easier and more cost-effective.
  • Designed to support rigorous applications.
Catalog No. 50-005-08344 Supplier MSC Supplier No. 55873640
The radial seal air filter element is designed to protect heavy-duty equipment by preventing airborne dirt and other contaminants from reaching the combustion chamber, ensuring proper equipment function.
  • Radial seal design utilizes a urethane compound to form molded ends and bond the media pack.
  • The design creates a strong, yet flexible package.
  • Heavy-duty radial seal gasket makes servicing easier and more cost-effective.
  • Designed to support rigorous applications.