Learn More
Grainger Baling Wire Coil Bare Wire
Catalog No. 502766873
Encompass_Preferred
The Malin Company Baling Wire is bare steel wire used for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13 gauge. The wire comes in coils that are approximately 1250 feet long and weigh around 55 pounds. This domestic baling wire is rust resistant and can be used with hand tie or automatic balers. It meets ASTM industry standards. The wire is an economical choice for baling applications on farms, ranches, and feedlots.
Catalog No. 50-276-6873 Supplier Grainger Supplier No. 16X998
The Malin Company Baling Wire is bare steel wire used for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13 gauge. The wire comes in coils that are approximately 1250 feet long and weigh around 55 pounds. This domestic baling wire is rust resistant and can be used with hand tie or automatic balers. It meets ASTM industry standards. The wire is an economical choice for baling applications on farms, ranches, and feedlots.