Learn More
Grainger Baling Wire Coil Bare Wire
Catalog No. 502767809
Encompass_Preferred
The Malin Company Baling Wire is a bare steel wire used for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13 gauge. The wire comes in 5 lb spools and has a smooth finish. It meets ASTM A510 standards. The wire's diameter measures 0.092 inches. It is manufactured in the USA by Malin Company, which has over 50 years of experience making quality baling wire. This is an economical wire solution for baling applications.
Catalog No. 50-276-7809 Supplier Grainger Supplier No. 16X984
The Malin Company Baling Wire is a bare steel wire used for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13 gauge. The wire comes in 5 lb spools and has a smooth finish. It meets ASTM A510 standards. The wire's diameter measures 0.092 inches. It is manufactured in the USA by Malin Company, which has over 50 years of experience making quality baling wire. This is an economical wire solution for baling applications.