Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502766819
Encompass_Preferred
The Grainger 16Y002 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This 10 lb stainless steel cylinder extinguisher uses Novec 1230 fluid which leaves no residue and is safe for use around sensitive electronics. It has a discharge range of 10 ft - 15 ft and can operate between -4°F to 120°F. The extinguisher is UL rated 2-A:10-B:C and comes with a 5 year limited warranty from the manufacturer.
Catalog No. 50-276-6819 Supplier Grainger Supplier No. 16Y002
The Grainger 16Y002 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This 10 lb stainless steel cylinder extinguisher uses Novec 1230 fluid which leaves no residue and is safe for use around sensitive electronics. It has a discharge range of 10 ft - 15 ft and can operate between -4°F to 120°F. The extinguisher is UL rated 2-A:10-B:C and comes with a 5 year limited warranty from the manufacturer.