Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502766893
Encompass_Preferred
The Malin Company Baling Wire is a bare steel wire used for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13 gauge. The wire comes in coils that are 350 feet long and weigh approximately 35 pounds. The wire is pliable and easy to work with. It meets ASTM A510 standards for baling wire. The wire is made in the USA by Malin Company, which has over 50 years of experience manufacturing quality baling wire and accessories. This is a versatile, heavy-duty baling wire suitable for agricultural baling applications.
Catalog No. 50-276-6893 Supplier Grainger Supplier No. 16Y008
The Malin Company Baling Wire is a bare steel wire used for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13 gauge. The wire comes in coils that are 350 feet long and weigh approximately 35 pounds. The wire is pliable and easy to work with. It meets ASTM A510 standards for baling wire. The wire is made in the USA by Malin Company, which has over 50 years of experience manufacturing quality baling wire and accessories. This is a versatile, heavy-duty baling wire suitable for agricultural baling applications.