Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767832
Encompass_Preferred
The Malin Company Baling Wire is bare steel wire used for baling hay, straw, and other agricultural materials. It is available in 13 gauge high tensile steel and comes in 1200 foot rolls. The wire has a minimum tensile strength of 60,000 psi. It can be used with hand tie baling machines and automatic wire tie balers. The wire meets ASTM A510 standards.
Catalog No. 50-276-7832 Supplier Grainger Supplier No. 16X989
The Malin Company Baling Wire is bare steel wire used for baling hay, straw, and other agricultural materials. It is available in 13 gauge high tensile steel and comes in 1200 foot rolls. The wire has a minimum tensile strength of 60,000 psi. It can be used with hand tie baling machines and automatic wire tie balers. The wire meets ASTM A510 standards.