Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767952
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13, 16, and 18 gauge sizes in lengths of 2000 feet. The wire comes in coils with inner diameters of 14, 16, or 18 inches. This general use baling wire helps make tight, weather-resistant bales. It meets ASTM A510 standards for steel wire rods. The wire is manufactured in the USA by Malin Company, which has over 50 years of experience making quality baling wire and accessories.
Catalog No. 50-276-7952 Supplier Grainger Supplier No. 16Y015
The Malin Company Baling Wire is a bare wire made of high carbon steel for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13, 16, and 18 gauge sizes in lengths of 2000 feet. The wire comes in coils with inner diameters of 14, 16, or 18 inches. This general use baling wire helps make tight, weather-resistant bales. It meets ASTM A510 standards for steel wire rods. The wire is manufactured in the USA by Malin Company, which has over 50 years of experience making quality baling wire and accessories.