Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767973
Encompass_Preferred
The Malin Company Baling Wire is bare steel wire used for baling hay, straw, and other agricultural materials. It is available in 13 gauge high tensile steel and comes in 1200 foot coils. The wire has a minimum tensile strength of 60,000 psi. It can be used with hand tie balers or automatic wire tie balers. The 13 gauge wire is 0.092 inches in diameter. This general use baling wire helps make tight, durable bales.
Catalog No. 50-276-7973 Supplier Grainger Supplier No. 16X991
The Malin Company Baling Wire is bare steel wire used for baling hay, straw, and other agricultural materials. It is available in 13 gauge high tensile steel and comes in 1200 foot coils. The wire has a minimum tensile strength of 60,000 psi. It can be used with hand tie balers or automatic wire tie balers. The 13 gauge wire is 0.092 inches in diameter. This general use baling wire helps make tight, durable bales.