Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767998
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, 18 gauge sizes in 1250 ft coils. The wire is pliable and sold in bulk for baling hay, straw, cotton and other agricultural products. The wire allows tight uniform bales that are easily handled and transported. It meets ASTM A510 standards for baling wire. The wire is sold through Grainger in the United States.
Catalog No. 50-276-7998 Supplier Grainger Supplier No. 16Y019
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, 18 gauge sizes in 1250 ft coils. The wire is pliable and sold in bulk for baling hay, straw, cotton and other agricultural products. The wire allows tight uniform bales that are easily handled and transported. It meets ASTM A510 standards for baling wire. The wire is sold through Grainger in the United States.