Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502768231
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes and comes in coils of 1250 feet. The wire is designed for baling hay, straw, and other agricultural materials. It features smooth wire that resists kinking and breaking. The wire's high tensile strength allows tight and secure bales. It can be used with both hand tie and automatic balers. This is a quality baling wire from Malin Company suitable for agricultural baling needs.
Catalog No. 50-276-8231 Supplier Grainger Supplier No. 16Y003
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes and comes in coils of 1250 feet. The wire is designed for baling hay, straw, and other agricultural materials. It features smooth wire that resists kinking and breaking. The wire's high tensile strength allows tight and secure bales. It can be used with both hand tie and automatic balers. This is a quality baling wire from Malin Company suitable for agricultural baling needs.