Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502766903
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13, 16, and 18 gauge sizes in 1200 ft coils. The wire is pliable and sold in bulk quantity for farm and ranch use. It meets ASTM A510 standards for baling wire. The wire helps make tight, durable bales that are easy to handle and transport.
Catalog No. 50-276-6903 Supplier Grainger Supplier No. 16Y028
Add to Cart
Add to Cart
The Malin Company Baling Wire is a bare wire made of high carbon steel for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13, 16, and 18 gauge sizes in 1200 ft coils. The wire is pliable and sold in bulk quantity for farm and ranch use. It meets ASTM A510 standards for baling wire. The wire helps make tight, durable bales that are easy to handle and transport.