Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502766574
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 16 gauge with a diameter of 0.0508 inches. The wire comes in coils of approximately 1320 feet and has many agricultural uses including baling hay, straw, and cotton. The wire is pliable and sold in bulk quantities. It can be used with hand baling tools or commercial balers to efficiently bundle agricultural materials for storage and transport. The high tensile strength makes it sturdy for heavy baling applications.
Catalog No. 50-276-6574 Supplier Grainger Supplier No. 16X999
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 16 gauge with a diameter of 0.0508 inches. The wire comes in coils of approximately 1320 feet and has many agricultural uses including baling hay, straw, and cotton. The wire is pliable and sold in bulk quantities. It can be used with hand baling tools or commercial balers to efficiently bundle agricultural materials for storage and transport. The high tensile strength makes it sturdy for heavy baling applications.