Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767856
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13, 16, and 18 gauge sizes in 1200 ft coils. The wire is pliable and sold in bulk quantity for farm and ranch use. It meets ASTM A510 standards for baling wire. The wire coils weigh approximately 55-85 lbs depending on gauge size.
Catalog No. 50-276-7856 Supplier Grainger Supplier No. 16Y027
The Malin Company Baling Wire is a bare wire made of high carbon steel for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13, 16, and 18 gauge sizes in 1200 ft coils. The wire is pliable and sold in bulk quantity for farm and ranch use. It meets ASTM A510 standards for baling wire. The wire coils weigh approximately 55-85 lbs depending on gauge size.