Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767859
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13, 16, and 18 gauge sizes in 1200 ft coils. The wire is pliable and sold in bulk quantity for farm and ranch use. It meets ASTM A510 standards for baling wire. The wire helps bind bales tightly for storage and transport. It is a cost-effective baling solution for agricultural operations.
Catalog No. 50-276-7859 Supplier Grainger Supplier No. 16Y020
The Malin Company Baling Wire is a bare wire made of high carbon steel for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13, 16, and 18 gauge sizes in 1200 ft coils. The wire is pliable and sold in bulk quantity for farm and ranch use. It meets ASTM A510 standards for baling wire. The wire helps bind bales tightly for storage and transport. It is a cost-effective baling solution for agricultural operations.