Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767925
Encompass_Preferred
The Malin Company Baling Wire is bare wire made of high carbon steel with a tensile strength of 60,000-70,000 psi. It comes in 16 gauge thickness and is available in rolls of 1,200 ft. The wire can be used for baling hay, straw, paper, cotton and more. It helps secure bales tightly. The wire features a smooth finish and consistent quality. It meets ASTM A510 standards. The wire works with most baling machines and hand balers. It offers durability and reliable binding strength.
Catalog No. 50-276-7925 Supplier Grainger Supplier No. 16X987
The Malin Company Baling Wire is bare wire made of high carbon steel with a tensile strength of 60,000-70,000 psi. It comes in 16 gauge thickness and is available in rolls of 1,200 ft. The wire can be used for baling hay, straw, paper, cotton and more. It helps secure bales tightly. The wire features a smooth finish and consistent quality. It meets ASTM A510 standards. The wire works with most baling machines and hand balers. It offers durability and reliable binding strength.