Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767927
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes in coils of 1250 feet. The wire is designed for baling hay, straw, and other agricultural materials. It features smooth wire that resists kinking and tangling. The wire meets ASTM A510 standards. It can be used with manual, semi-automatic, or automatic balers. The wire is made in the USA.
Catalog No. 50-276-7927 Supplier Grainger Supplier No. 16Y032
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes in coils of 1250 feet. The wire is designed for baling hay, straw, and other agricultural materials. It features smooth wire that resists kinking and tangling. The wire meets ASTM A510 standards. It can be used with manual, semi-automatic, or automatic balers. The wire is made in the USA.