Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502766887
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes in coils of 1250 feet. The wire is designed for baling hay, straw, and other agricultural materials. It features smooth wire that resists kinking and tangling. The wire allows tight, secure bales that hold their shape for transport and storage. It can be used with both hand tie and automatic balers. The wire is sold by the coil and priced economically for high volume agricultural use.
Catalog No. 50-276-6887 Supplier Grainger Supplier No. 16Y041
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes in coils of 1250 feet. The wire is designed for baling hay, straw, and other agricultural materials. It features smooth wire that resists kinking and tangling. The wire allows tight, secure bales that hold their shape for transport and storage. It can be used with both hand tie and automatic balers. The wire is sold by the coil and priced economically for high volume agricultural use.