Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502766889
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes and comes in coils of 1250 feet. The wire can be used for baling hay, straw, and other agricultural products. It features smooth wire that will not damage bales. The wire is pliable and easy to work with. It provides economical binding power. The wire meets ASTM A510 standards.
Catalog No. 50-276-6889 Supplier Grainger Supplier No. 16Y006
Add to Cart
Add to Cart
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes and comes in coils of 1250 feet. The wire can be used for baling hay, straw, and other agricultural products. It features smooth wire that will not damage bales. The wire is pliable and easy to work with. It provides economical binding power. The wire meets ASTM A510 standards.