Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502767861
Encompass_Preferred
The Malin Company Baling Wire is a bare steel wire used for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13 gauge. The wire comes in coils that are approximately 1250 feet long and weigh around 55 pounds. This domestic wire is rust resistant and can be used with hand tie or automatic baling machines. The wire meets ASTM A510 standards.
Catalog No. 50-276-7861 Supplier Grainger Supplier No. 16Y012
The Malin Company Baling Wire is a bare steel wire used for baling hay, straw, and other agricultural materials. It has high tensile strength and is available in 13 gauge. The wire comes in coils that are approximately 1250 feet long and weigh around 55 pounds. This domestic wire is rust resistant and can be used with hand tie or automatic baling machines. The wire meets ASTM A510 standards.