Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502766578
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It comes in 13 gauge thickness and is available in rolls of 1,200 feet. The wire can be used for baling hay, straw, and other agricultural materials. It features a smooth finish and consistent diameter throughout the coil. The wire is pliable and easy to work with by hand or with baling equipment. It provides the strength needed to tightly bale agricultural materials for storage and transportation. This is a quality baling wire suitable for farm and ranch use.
Catalog No. 50-276-6578 Supplier Grainger Supplier No. 16Y039
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It comes in 13 gauge thickness and is available in rolls of 1,200 feet. The wire can be used for baling hay, straw, and other agricultural materials. It features a smooth finish and consistent diameter throughout the coil. The wire is pliable and easy to work with by hand or with baling equipment. It provides the strength needed to tightly bale agricultural materials for storage and transportation. This is a quality baling wire suitable for farm and ranch use.