Learn More
Grainger Baling Wire Spool Bare Wire
Catalog No. 502766580
Encompass_Preferred
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes in coils of 1250 feet. The wire is designed for baling hay, straw, and other agricultural materials. It features smooth wire that resists kinking and tangling. The wire's high tensile strength allows tight, durable bales. It can be used with both hand tie and automatic balers. This is a quality baling wire from Malin Company suitable for agricultural baling applications.
Catalog No. 50-276-6580 Supplier Grainger Supplier No. 16Y022
The Malin Company Baling Wire is a bare wire made of high carbon steel with a tensile strength of 60,000 to 70,000 psi. It is available in 13, 16, and 18 gauge sizes in coils of 1250 feet. The wire is designed for baling hay, straw, and other agricultural materials. It features smooth wire that resists kinking and tangling. The wire's high tensile strength allows tight, durable bales. It can be used with both hand tie and automatic balers. This is a quality baling wire from Malin Company suitable for agricultural baling applications.