Learn More
Medchemexpress LLC Balixafortide TFA (POL6326 TFA) | 1051366-32-5 | 99.9% | 1978.16 | 25 MG
SDP
Catalog No. 5000426794
Encompass_Preferred
Balixafortide TFA (POL6326 TFA) is a potent, selective, and well-tolerated peptidic CXCR4 antagonist with an IC50 less than 10 nM. It demonstrates significant selectivity for CXCR4, approximately 1000-fold, over various other receptors including CXCR7. This compound effectively blocks β-arrestin recruitment and calcium flux with IC50s less than 10 nM, functioning as a potent hematopoietic stem and progenitor cell (HSPC) mobilizing agent with observed anti-cancer effects.
  • Potent and selective CXCR4 antagonist
  • Demonstrates high selectivity over other receptors like CXCR7
  • Blocks β-arrestin recruitment and calcium flux
  • Functions as a potent hematopoietic stem and progenitor cell (HSPC) mobilizing agent
  • Exhibits anti-cancer effects
  • Optimized for favorable mouse ADME properties
Catalog No. 50-004-26794 Supplier Medchemexpress LLC Supplier No. 25MGHYP1682A
Balixafortide TFA (POL6326 TFA) is a potent, selective, and well-tolerated peptidic CXCR4 antagonist with an IC50 less than 10 nM. It demonstrates significant selectivity for CXCR4, approximately 1000-fold, over various other receptors including CXCR7. This compound effectively blocks β-arrestin recruitment and calcium flux with IC50s less than 10 nM, functioning as a potent hematopoietic stem and progenitor cell (HSPC) mobilizing agent with observed anti-cancer effects.
  • Potent and selective CXCR4 antagonist
  • Demonstrates high selectivity over other receptors like CXCR7
  • Blocks β-arrestin recruitment and calcium flux
  • Functions as a potent hematopoietic stem and progenitor cell (HSPC) mobilizing agent
  • Exhibits anti-cancer effects
  • Optimized for favorable mouse ADME properties