Learn More
Anton Paar Set of 5 tungsten carbide balls 15 mm
Catalog No. NC1818293
Encompass_Preferred
Grinding ball with 15 mm diameter made of tungsten carbide for use with grinding jars made of tungsten carbide.Suitable for use with Anton Paar BM500 Ball Mill NC1818284 and Grinding Jar NC1818291
Catalog No. NC1818293 Supplier Anton Paar Supplier No. 179715
Grinding ball with 15 mm diameter made of tungsten carbide for use with grinding jars made of tungsten carbide.Suitable for use with Anton Paar BM500 Ball Mill NC1818284 and Grinding Jar NC1818291