Learn More
Grainger BALL BEARING,20MM BORE, ALLOY
Supplier:  Grainger 23Y213
Encompass_Preferred
SKF Radial Ball Bearing: 6204, Open, 20 mm Bore, 47 mm OD, 14 mm Wd, Alloy Steel Ring
Catalog No. NC2293526