Learn More
World Precision Instrument Ball Bearing Boom Stand
SDP
Catalog No. 502521352
Encompass
  • Dual Arm Ball Bearing construction increases stability and allows smoother horizontal movements.
  • Length of arm is 40 cm
  • Focus Mount must be purchased separately
Catalog No. 50-252-1352 Supplier World Precision Instrument Supplier No. 502005
  • Dual Arm Ball Bearing construction increases stability and allows smoother horizontal movements.
  • Length of arm is 40 cm
  • Focus Mount must be purchased separately