Learn More
MSC AMPG Ball Contact Point, Carbide, 0.04 in. x 1/2 in., 1 ea.
Catalog No. 5000514841
Encompass
This ball contact point is designed for high-precision measuring and marking applications. Constructed from durable carbide, it provides exceptional hardness and wear resistance suitable for demanding industrial environments including aerospace, automotive, and medical device manufacturing.
  • Carbide construction ensures longevity and superior wear resistance
  • Features a 0.04 in. ball diameter and 1/2 in. total length
  • Includes a 4-48 thread for secure attachment
  • Designed for high-precision measurement tasks
  • Ideal for use in metalworking and machining industries
Catalog No. 50-005-14841 Supplier MSC Supplier No. 81548349
This ball contact point is designed for high-precision measuring and marking applications. Constructed from durable carbide, it provides exceptional hardness and wear resistance suitable for demanding industrial environments including aerospace, automotive, and medical device manufacturing.
  • Carbide construction ensures longevity and superior wear resistance
  • Features a 0.04 in. ball diameter and 1/2 in. total length
  • Includes a 4-48 thread for secure attachment
  • Designed for high-precision measurement tasks
  • Ideal for use in metalworking and machining industries