Learn More
Grainger Ball End Mill Single End 0.20mm Carbide
Catalog No. 502751511
Encompass_Preferred
The Micro 100 14M522 is a 1/4" diameter, 2 flute, ball end mill made of solid carbide with a TiAlN coating. It has a 1/4" shank diameter and a 1-1/2" overall length. The ball end has a 0.0312" radius. This end mill is designed for general purpose milling in steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. Its TiAlN coating provides heat and wear resistance. The Micro 100 14M522 ball end mill is made in the USA.
Catalog No. 50-275-1511 Supplier Grainger Supplier No. 14M522
The Micro 100 14M522 is a 1/4" diameter, 2 flute, ball end mill made of solid carbide with a TiAlN coating. It has a 1/4" shank diameter and a 1-1/2" overall length. The ball end has a 0.0312" radius. This end mill is designed for general purpose milling in steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. Its TiAlN coating provides heat and wear resistance. The Micro 100 14M522 ball end mill is made in the USA.