Learn More
Grainger Ball End Mill Single End 0.30mm Carbide
Catalog No. 502750252
Encompass_Preferred
The Grainger Approved Ball End Mill Single End is a high-speed steel end mill with a ball end for milling contoured surfaces. It has a 1/4" shank diameter, 1/2" overall length, and a 1/4" cutting diameter. The end mill has a TiN coating for increased hardness and lubricity. It is designed for general purpose milling in most material types up to 45 HRC hardness. The end mill provides accurate results for profiling, contouring, slotting, and pocketing applications.
Catalog No. 50-275-0252 Supplier Grainger Supplier No. 14K319
The Grainger Approved Ball End Mill Single End is a high-speed steel end mill with a ball end for milling contoured surfaces. It has a 1/4" shank diameter, 1/2" overall length, and a 1/4" cutting diameter. The end mill has a TiN coating for increased hardness and lubricity. It is designed for general purpose milling in most material types up to 45 HRC hardness. The end mill provides accurate results for profiling, contouring, slotting, and pocketing applications.