Learn More
Grainger Ball End Mill Single End 0.40mm Carbide
Catalog No. 502749986
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, a 1/2" cutting diameter, and a 1-1/2" overall length. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in materials like aluminum, brass, bronze, and copper. The end mill can be run at speeds up to 11,000 RPM with feed rates up to 80 IPM. It is made in the USA by Ball, a manufacturer of precision cutting tools.
Catalog No. 50-274-9986 Supplier Grainger Supplier No. 14K324
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, a 1/2" cutting diameter, and a 1-1/2" overall length. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in materials like aluminum, brass, bronze, and copper. The end mill can be run at speeds up to 11,000 RPM with feed rates up to 80 IPM. It is made in the USA by Ball, a manufacturer of precision cutting tools.