Learn More
Grainger Ball End Mill Single End 0.40mm Carbide
Catalog No. 502749987
Encompass_Preferred
The Micro 100 Ball End Mill Carbide is a 2-flute ball end mill made of solid carbide with a TiAlN coating. It has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. This 2-flute design provides good chip ejection and finish. The TiAlN coating improves tool life and heat resistance. It can machine steels, stainless steels, cast iron, hardened steels, titanium, and nickel alloys. It has a center cutting design for plunging capability. Recommended speeds are 18,000-22,000 RPM with .001-.004" per tooth feed rates. It is well-suited for complex 3D contouring and profiling with its ball end geometry.
Catalog No. 50-274-9987 Supplier Grainger Supplier No. 14K323
The Micro 100 Ball End Mill Carbide is a 2-flute ball end mill made of solid carbide with a TiAlN coating. It has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. This 2-flute design provides good chip ejection and finish. The TiAlN coating improves tool life and heat resistance. It can machine steels, stainless steels, cast iron, hardened steels, titanium, and nickel alloys. It has a center cutting design for plunging capability. Recommended speeds are 18,000-22,000 RPM with .001-.004" per tooth feed rates. It is well-suited for complex 3D contouring and profiling with its ball end geometry.