Learn More
Grainger Ball End Mill Single End 0.40mm Carbide
Catalog No. 502750044
Encompass_Preferred
The Micro 100 Ball End Mill Carbide is a 2-flute ball end mill made of solid carbide with a TiAlN coating. It has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. The ball end geometry allows milling of 3D contoured surfaces. This end mill is designed for general purpose milling in most material groups like steel, stainless steel, cast iron, aluminum, and plastics. It provides good chip evacuation and can mill up to 50% radial engagement and 25% axial engagement. The TiAlN coating reduces heat and friction for faster cutting. The end mill has center cutting geometry for plunge cuts.
Catalog No. 50-275-0044 Supplier Grainger Supplier No. 14K325
The Micro 100 Ball End Mill Carbide is a 2-flute ball end mill made of solid carbide with a TiAlN coating. It has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. The ball end geometry allows milling of 3D contoured surfaces. This end mill is designed for general purpose milling in most material groups like steel, stainless steel, cast iron, aluminum, and plastics. It provides good chip evacuation and can mill up to 50% radial engagement and 25% axial engagement. The TiAlN coating reduces heat and friction for faster cutting. The end mill has center cutting geometry for plunge cuts.