Learn More
Grainger Ball End Mill Single End 0.40mm Carbide
Catalog No. 502750253
Encompass_Preferred
The Micro 100 Ball End Mill Carbide is a high-performance end mill for milling a variety of materials. It has a ball end geometry for milling 3D contoured shapes and is made of solid carbide for exceptional wear resistance and long tool life. This 2-flute end mill has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2" for different milling applications. The TiAlN coating provides a high hardness surface and thermal barrier. Overall, this is a versatile end mill suitable for high-speed machining of steel, stainless steel, cast iron, aluminum, and other non-ferrous metals.
Catalog No. 50-275-0253 Supplier Grainger Supplier No. 14K322
The Micro 100 Ball End Mill Carbide is a high-performance end mill for milling a variety of materials. It has a ball end geometry for milling 3D contoured shapes and is made of solid carbide for exceptional wear resistance and long tool life. This 2-flute end mill has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2" for different milling applications. The TiAlN coating provides a high hardness surface and thermal barrier. Overall, this is a versatile end mill suitable for high-speed machining of steel, stainless steel, cast iron, aluminum, and other non-ferrous metals.