Learn More
Grainger Ball End Mill Single End 0.50mm Carbide
Catalog No. 502751201
Encompass_Preferred
The Grainger 14K329 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.6/0.8 amps and is ideal for driving pumps, fans, and shop equipment. This electric motor would be suitable for industrial, commercial, or residential applications that require up to 3/4 horsepower.
Catalog No. 50-275-1201 Supplier Grainger Supplier No. 14K329
The Grainger 14K329 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.6/0.8 amps and is ideal for driving pumps, fans, and shop equipment. This electric motor would be suitable for industrial, commercial, or residential applications that require up to 3/4 horsepower.