Learn More
Grainger Ball End Mill Single End 0.60mm Carbide
Catalog No. 502751202
Encompass_Preferred
The Grainger Approved Ball End Mill is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, 1/2" cutting diameter, 2" overall length, and 2 flute design. This end mill is suitable for slotting, contouring and profiling in most work materials. It can be run at speeds up to 5500 RPM with conventional high speed steel tooling. The ball nose produces a smooth milling action that makes it ideal for 3D contour work and complex surface slotting.
Catalog No. 50-275-1202 Supplier Grainger Supplier No. 14K331
The Grainger Approved Ball End Mill is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, 1/2" cutting diameter, 2" overall length, and 2 flute design. This end mill is suitable for slotting, contouring and profiling in most work materials. It can be run at speeds up to 5500 RPM with conventional high speed steel tooling. The ball nose produces a smooth milling action that makes it ideal for 3D contour work and complex surface slotting.