Learn More
Grainger Ball End Mill Single End 1.00mm Carbide
Catalog No. 502749984
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 1/2" cutting diameter, and a 2" overall length. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in steels, stainless steels, cast iron, hardened steels, titanium, and other materials. The end mill provides accurate radii for 3D contouring and can be run at speeds up to 2000 SFM. It is made in the USA by Ball, a manufacturer of precision cutting tools.
Catalog No. 50-274-9984 Supplier Grainger Supplier No. 14K351
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 1/2" cutting diameter, and a 2" overall length. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in steels, stainless steels, cast iron, hardened steels, titanium, and other materials. The end mill provides accurate radii for 3D contouring and can be run at speeds up to 2000 SFM. It is made in the USA by Ball, a manufacturer of precision cutting tools.