Learn More
Grainger Ball End Mill Single End 1.00mm Carbide
Catalog No. 502750248
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in materials like steel, stainless steel, cast iron, aluminum, copper, and plastics. The ball nose geometry allows milling of 3D contoured shapes.
Catalog No. 50-275-0248 Supplier Grainger Supplier No. 14K355
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in materials like steel, stainless steel, cast iron, aluminum, copper, and plastics. The ball nose geometry allows milling of 3D contoured shapes.