Learn More
Grainger Ball End Mill Single End 1.00mm Carbide
Supplier:  Grainger 14K350
Encompass_Preferred
The Grainger 14K350 is a 14 gauge, type K thermocouple with a male mini-connector on one end and an exposed thermocouple junction on the other end. It has a temperature range of -454 to 2282°F (-270 to 1250°C) and is made of Chromel and Alumel conductors. This 36 inch long thermocouple features abrasion resistance, flexibility, and chemical stability. It can be used with controllers or transmitters that accept type K mini-connectors for temperature measurement in industrial, commercial, and food service applications. The 14K350 is RoHS compliant.
Catalog No. 50-275-0250