Learn More
Grainger Ball End Mill Single End 1.00mm Carbide
Catalog No. 502751238
Encompass_Preferred
The Grainger Approved 1.00mm Carbide Ball End Mill Single End is a milling tool with a 1.00mm diameter ball end made of solid carbide for high speed machining in aluminum, brass, copper, graphite, plastics, and wood. It has a shank diameter of 3mm, overall length of 20mm, and 2 flutes. This single end mill for light milling applications features TiAIN coating for heat and wear resistance. It has a right-hand spiral flute direction.
Catalog No. 50-275-1238 Supplier Grainger Supplier No. 14K358
The Grainger Approved 1.00mm Carbide Ball End Mill Single End is a milling tool with a 1.00mm diameter ball end made of solid carbide for high speed machining in aluminum, brass, copper, graphite, plastics, and wood. It has a shank diameter of 3mm, overall length of 20mm, and 2 flutes. This single end mill for light milling applications features TiAIN coating for heat and wear resistance. It has a right-hand spiral flute direction.