Learn More
Grainger Ball End Mill Single End 1.00mm Carbide
Catalog No. 502750041
Encompass_Preferred
The Grainger 14K357 is a 14 gauge, type 304 stainless steel wire cloth with a 2 mesh and 0.045 inch wire diameter. It has an open area of 36% and comes in a 48 inch width. The stainless steel construction makes it durable and corrosion resistant. With a fine mesh, this product can be used in various filtration, screening, and sieving applications. It is made in the USA.
Catalog No. 50-275-0041 Supplier Grainger Supplier No. 14K357
The Grainger 14K357 is a 14 gauge, type 304 stainless steel wire cloth with a 2 mesh and 0.045 inch wire diameter. It has an open area of 36% and comes in a 48 inch width. The stainless steel construction makes it durable and corrosion resistant. With a fine mesh, this product can be used in various filtration, screening, and sieving applications. It is made in the USA.