Learn More
Grainger Ball End Mill Single End 1.50mm Carbide
Catalog No. 502750535
Encompass_Preferred
The WIDIA HANITA Ball End Mill is a solid carbide end mill with a ball nose end. It has a cutting diameter of 1/4", shank diameter of 1/4", overall length of 1-1/2", and 2 flutes. The end mill features a TiAlN coating for increased tool life and heat resistance. It is designed for high speed machining in a variety of materials including hardened steels, titanium alloys, nickel based alloys, and other exotics. This end mill is made in Israel by WIDIA, part of the Kennametal group, a leading manufacturer of cutting tools.
Catalog No. 50-275-0535 Supplier Grainger Supplier No. 14K372
The WIDIA HANITA Ball End Mill is a solid carbide end mill with a ball nose end. It has a cutting diameter of 1/4", shank diameter of 1/4", overall length of 1-1/2", and 2 flutes. The end mill features a TiAlN coating for increased tool life and heat resistance. It is designed for high speed machining in a variety of materials including hardened steels, titanium alloys, nickel based alloys, and other exotics. This end mill is made in Israel by WIDIA, part of the Kennametal group, a leading manufacturer of cutting tools.