Learn More
Grainger Ball End Mill Single End 10.00mm Carbide
Catalog No. 502749505
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end for milling contoured surfaces. It has a shank diameter of 1/2", an overall length of 4", and a ball diameter of 1/2". The end mill has a TiAlN coating for increased tool life and heat resistance. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and graphite. The end mill is designed for high speed machining with recommended speeds up to 11,000 RPM and feed rates up to 80 IPM. It can be used for slotting, side milling and profiling operations. The end mill provides accurate results and smooth finishes.
Catalog No. 50-274-9505 Supplier Grainger Supplier No. 14K809
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end for milling contoured surfaces. It has a shank diameter of 1/2", an overall length of 4", and a ball diameter of 1/2". The end mill has a TiAlN coating for increased tool life and heat resistance. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and graphite. The end mill is designed for high speed machining with recommended speeds up to 11,000 RPM and feed rates up to 80 IPM. It can be used for slotting, side milling and profiling operations. The end mill provides accurate results and smooth finishes.