Learn More
Grainger Ball End Mill Single End 10.00mm Carbide
Catalog No. 502751296
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, 1/2" overall length, and a 1/4" cutting diameter. The end mill has 2 flutes and a right hand spiral. It is designed for general purpose milling in materials like aluminum, brass, bronze, and plastics. The ball nose end allows milling of 3D contoured surfaces.
Catalog No. 50-275-1296 Supplier Grainger Supplier No. 14K810
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, 1/2" overall length, and a 1/4" cutting diameter. The end mill has 2 flutes and a right hand spiral. It is designed for general purpose milling in materials like aluminum, brass, bronze, and plastics. The ball nose end allows milling of 3D contoured surfaces.