Learn More
Grainger Approved Vendor Ball End Mill, Carbide, AlTiN Coated, 11/64 in. Diameter, 1 ea.
Catalog No. 502782769
Encompass_Preferred
This ball end mill is manufactured from sub-micron grain carbide with 10% cobalt and features an aluminum titanium nitride (AlTiN) coating for high-performance machining. Designed with two flutes and a center-cutting geometry, it is suitable for a wide range of materials including steel, stainless steel, and aluminum alloys. Its ball nose design provides versatility for three-axis contouring, slotting, pocketing, and plunging operations.
  • Carbide construction with 10% cobalt provides durability and excellent heat resistance
  • AlTiN coating is ideal for dry machining applications
  • Center cutting capability allows for effective plunging
  • Suitable for profiling, slotting, and contour finishing
  • Features a 3/16 in. shank diameter and 11/64 in. mill diameter
Catalog No. 50-278-2769 Supplier Grainger Supplier No. 19LM66
This ball end mill is manufactured from sub-micron grain carbide with 10% cobalt and features an aluminum titanium nitride (AlTiN) coating for high-performance machining. Designed with two flutes and a center-cutting geometry, it is suitable for a wide range of materials including steel, stainless steel, and aluminum alloys. Its ball nose design provides versatility for three-axis contouring, slotting, pocketing, and plunging operations.
  • Carbide construction with 10% cobalt provides durability and excellent heat resistance
  • AlTiN coating is ideal for dry machining applications
  • Center cutting capability allows for effective plunging
  • Suitable for profiling, slotting, and contour finishing
  • Features a 3/16 in. shank diameter and 11/64 in. mill diameter