Learn More
Grainger Ball End Mill Single End 12.00mm Carbide
Supplier:  Grainger 14M228
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for general purpose milling in materials like steel, stainless steel, cast iron, aluminum, copper, and plastics. The ball nose geometry allows milling of 3D contoured surfaces. This is a quality end mill made in the USA.
Catalog No. 50-274-9893